Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slco2a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033314.3 | NP_201571.2 | 1, 2 | 235 | ||
GenBank mRNA | AF323958.1 | - | 1, 2 | 104 | ||
AK087350.1 | - | 1, 2 | 238 | |||
AK166724.1 | - | 1, 2 | 236 | |||
AK169706.1 | - | 1, 2 | 238 | |||
BC035200.1 | - | 225 | 3158233 |