Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tfcp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289603.1 | NP_001276532.1 | 3, 4 | 416 | ||
NM_033476.3 | NP_258437.1 | 3, 4 | 416 | |||
GenBank mRNA | AK051891.1 | - | 2, 3 | 415 | ||
AK131989.1 | - | 2, 3 | 615 | |||
AK146503.1 | - | 3, 4 | 417 | |||
AK147898.1 | - | 2, 3 | 625 | |||
AK158682.1 | - | 3, 4 | 336 | |||
BC022131.1 | - | 3, 4 | 389 | 5098336 | ||
M84809.1 | - | 3, 4 | 355 |