Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif4h | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033561.1 | NP_291039.1 | 2 | 202 | ||
GenBank mRNA | AK045004.1 | - | 2 | 204 | ||
AK088818.1 | - | 2 | 214 | |||
AK122194.1 | - | 2 | 202 | |||
AK132034.1 | - | 2 | 222 | |||
AK135390.1 | - | 2 | 203 | |||
AK136999.1 | - | 2 | 189 | |||
AK149841.1 | - | 2 | 188 | |||
AK150088.1 | - | 2 | 211 | |||
AK166567.1 | - | 2 | 203 | |||
AK168872.1 | - | 2 | 221 | |||
AK169312.1 | - | 2 | 185 | |||
AK169353.1 | - | 2 | 203 | |||
AK169407.1 | - | 2 | 221 | |||
AK185835.1 | - | 1 | 39 | |||
AK194751.1 | - | 1 | 39 | |||
AK208946.1 | - | 1 | 39 | |||
AK211404.1 | - | 1 | 39 | |||
BC014796.1 | - | 2 | 216 | 3962104 |