Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nrg4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032002.2 | NP_114391.1 | 4 | 332 | ||
GenBank mRNA | AF083067.2 | - | 3 | 210 | ||
AK041077.1 | - | 5 | 364 | |||
AK080058.1 | - | 4 | 284 | |||
AK080089.1 | - | 4 | 310 | |||
AK080161.1 | - | 5 | 364 | |||
BC034839.1 | - | 7 | 625 | 3372942 |