Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gtpbp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032544.3 | NP_115933.2 | 4 | 1170 | ||
GenBank mRNA | AF361482.2 | - | 4 | 1170 | ||
AK046330.1 | - | 4 | 546 | |||
AK219310.1 | - | 1 | 66 | |||
BC055716.1 | - | 4 | 522 | |||
BC075712.1 | - | 4 | 530 | |||
BC092042.1 | - | 4 | 553 | |||
BC138560.1 | - | 4 | 579 | 8861580 | ||
BC145333.1 | - | 4 | 575 |