Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mbd6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033072.2 | NP_149063.2 | 13 | 3538 | ||
GenBank mRNA | AK018262.1 | - | 4 | 679 | ||
AK047331.1 | - | 3 | 563 | |||
AK158797.1 | - | 13 | 3656 | |||
BC019433.1 | - | 5 | 1039 | |||
BC038259.1 | - | 5 | 457 | |||
BC049833.1 | - | 5 | 1022 | |||
BC055445.1 | - | 5 | 995 | |||
BC059015.1 | - | 14 | 3321 | |||
BC083328.1 | - | 4 | 675 | |||
BC096027.1 | - | 10 | 3280 |