Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Actn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033268.4 | NP_150371.4 | 3 | 570 | ||
GenBank mRNA | AF248643.1 | - | 3 | 338 | ||
AK047768.1 | - | 3 | 571 | |||
AY036877.1 | - | 4 | 514 | |||
BC089579.1 | - | 3 | 570 | 6757718 |