Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Glce | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033320.4 | NP_201577.3 | 1 | 308 | ||
GenBank mRNA | AF325532.1 | - | 3 | 533 | ||
AF330049.1 | - | 1 | 542 | |||
BC094587.1 | - | 1 | 308 | 30618522 |