Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dgcr8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033324.2 | NP_201581.2 | 2 | 702 | ||
GenBank mRNA | AB086857.1 | - | 2 | 702 | ||
AF201683.1 | - | 1 | 356 | |||
AK048056.1 | - | 1 | 503 | |||
AK086233.1 | - | 2 | 859 | |||
BC051013.1 | - | 1 | 475 | |||
BC062919.1 | - | 2 | 722 | 6833280 | ||
BC066118.1 | - | 2 | 749 | 6400561 |