Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slmap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032008.3 | NP_114397.3 | 20, 21 | 2455 | ||
GenBank mRNA | AF304451.1 | - | 8, 9 | 1003 | ||
AK129403.1 | - | 20, 21 | 2455 | |||
AK141597.1 | - | 21, 22 | 3675 | |||
AK146685.1 | - | 8, 9 | 969 | |||
BC021457.1 | - | 10, 11 | 1218 | 5064534 |