Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kremen1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032396.3 | NP_115772.2 | 5 | 701 | ||
GenBank mRNA | AB059617.1 | - | 5 | 664 | ||
AK083565.1 | - | 3 | 309 | |||
AK141321.1 | - | 5 | 713 | |||
BC049771.1 | - | 5 | 625 | |||
BC082546.1 | - | 5 | 701 | 6855459 | ||
BC138464.1 | - | 5 | 709 | 8861484 | ||
BC138465.1 | - | 5 | 709 | 8861485 |