Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033037.3 | NP_149026.1 | 3 | 506 | ||
GenBank mRNA | AK004249.1 | - | 3 | 520 | ||
AK011032.1 | - | 3 | 520 | |||
AK149582.1 | - | 3 | 520 | |||
BC013638.1 | - | 3 | 506 | 4194939 | ||
BC020375.1 | - | 3 | 506 |