Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Inpp5e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290437.1 | NP_001277366.1 | 11 | 2397 | ||
NM_033134.3 | NP_149125.1 | 10 | 2728 | |||
GenBank mRNA | AF226683.1 | - | 10 | 2690 | ||
AK047245.1 | - | 8 | 2657 | |||
AK080075.1 | - | 10 | 2710 | |||
AK085439.1 | - | 11 | 2402 | |||
AK154097.1 | - | 10 | 2729 | |||
AK158135.1 | - | 10 | 2713 | |||
AK170174.1 | - | 10 | 2798 | |||
AK170786.1 | - | 10 | 2685 | |||
AK170812.1 | - | 9 | 2763 | |||
BC052717.1 | - | 10 | 2697 | 6837339 | ||
BC057566.1 | - | 10 | 2780 | |||
BC080295.1 | - | 10 | 2693 | 30354397 |