Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: P2rx5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033321.3 | NP_201578.2 | 12 | 1215 | ||
GenBank mRNA | AF333331.1 | - | 12 | 1221 | ||
AK134662.1 | - | 12 | 1175 | |||
BC138189.1 | - | 12 | 1215 | 8861208 | ||
BC145181.1 | - | 12 | 1215 | 9053732 | ||
JX570808.1 | - | 11 | 1478 |