Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cnnm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102471.1 | NP_001095941.1 | 5 | 2276 | ||
NM_033569.3 | NP_291047.2 | 5 | 2276 | |||
GenBank mRNA | AF216961.1 | - | 5 | 1588 | ||
AK032006.1 | - | 5 | 2235 | |||
AK053711.1 | - | 5 | 2294 | |||
AK159616.1 | - | 5 | 2282 | |||
BC038027.1 | - | 168 | ||||
BC052513.1 | - | 3 | 291 | |||
BC125033.1 | - | 5 | 1588 | |||
GQ225745.1 | - | 5 | 1575 |