Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcdhgb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033576.1 | NP_291054.1 | 1 | 1460 | ||
GenBank mRNA | AK083581.1 | - | 1 | 1631 | ||
AY013806.1 | - | 1 | 1460 | |||
BC138701.1 | - | 1 | 1556 | 8861723 |