Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcdhgb6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033578.3 | NP_291056.1 | 1 | 2438 | ||
GenBank mRNA | AY013808.1 | - | 1 | 2291 | ||
BC054556.1 | - | 1 | 2438 | 5702016 |