Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRYAA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000394.3 | NP_000385.1 | 2 | 370 | ||
XM_005261093.3 | XP_005261150.1 | 2 | 475 | |||
GenBank mRNA | BC113598.1 | - | 2 | 299 | 8322650 | |
BX118596.1 | - | 2 | 370 | |||
CR407691.1 | - | 2 | 278 | |||
KM220588.1 | - | 2 | 328 | |||
KM220590.1 | - | 2 | 335 | |||
KM220591.1 | - | 2 | 294 | |||
KM220592.1 | - | 2 | 336 | |||
U05569.1 | - | 2 | 345 | |||
U66584.1 | - | 2 | 278 | |||
X14789.1 | - | 1 | 1519 |
Gene Symbol: LOC102724652 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001314050.2 | NP_001300979.1 | 2 | 337 | ||
NM_001320719.1 | NP_001307648.1 | 2 | 263 | |||
GenBank mRNA | BC069528.1 | - | 2 | 328 | 7262133 | |
BQ640485.1 | - | 2 | 263 | |||
KM220589.1 | - | 2 | 333 |