Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Igsf9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145800.1 | NP_001139272.1 | 4 | 532 | ||
NM_033608.3 | NP_291086.2 | 4 | 598 | |||
GenBank mRNA | AF317839.1 | - | 4 | 598 | ||
AK053544.1 | - | 4 | 526 | |||
AK078536.1 | - | 4 | 526 | |||
AK145121.1 | - | 4 | 507 | |||
BC034594.1 | - | 4 | 514 | |||
BC040281.1 | - | 4 | 558 | 5364073 |