Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ophn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052976.3 | NP_443208.1 | 2 | 724 | ||
GenBank mRNA | AK028822.1 | - | 2 | 208 | ||
AK031419.1 | - | 2 | 717 | |||
AK032500.1 | - | 2 | 366 | |||
AK036038.1 | - | 2 | 725 | |||
AK039304.1 | - | 1 | 892 | |||
AK043239.1 | - | 2 | 725 | |||
AK046214.1 | - | 2 | 180 | |||
AK048153.1 | - | 2 | 725 | |||
AK077982.1 | - | 2 | 349 | |||
AK082551.1 | - | 2 | 364 | |||
AK087469.1 | - | 2 | 188 | |||
AK166392.1 | - | 2 | 728 | |||
BC004845.1 | - | 3 | 322 | 2650009 |