Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrp1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053011.2 | NP_443737.2 | 3, 4 | 1118 | ||
GenBank mRNA | AF270884.1 | - | 3, 4 | 1118 | ||
AK039425.1 | - | 3, 4 | 595 |