Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam126a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053090.2 | NP_444320.2 | 3, 4 | 379 | ||
GenBank mRNA | AK155209.1 | - | 3, 4 | 380 | ||
AK161265.1 | - | 3, 4 | 573 | |||
BC056381.1 | - | 3, 4 | 318 | 6852011 | ||
BC060692.1 | - | 3, 4 | 366 | 6410470 |