Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: G6PD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000402.4 | NP_000393.4 | 9 | 1116 | ||
NM_001042351.2 | NP_001035810.1 | 9 | 1005 | |||
GenBank mRNA | AK292304.1 | - | 9 | 942 | ||
AK302341.1 | - | 9 | 943 | |||
BC000337.2 | - | 9 | 931 | 2822640 | ||
M21248.1 | - | 8 | 892 | |||
X03674.1 | - | 9 | 1348 |