Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GALE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000403.3 | NP_000394.2 | 12 | 1469 | ||
NM_001008216.1 | NP_001008217.1 | 12 | 1404 | |||
NM_001127621.1 | NP_001121093.1 | 11 | 1445 | |||
GenBank mRNA | AK057302.1 | - | 10 | 1640 | ||
BC001273.1 | - | 11 | 1325 | 3459004 | ||
BC050685.2 | - | 12 | 1328 | 6161561 | ||
L41668.1 | - | 12 | 1327 |