Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000404.3 | NP_000395.2 | 11, 12 | 1281 | ||
NM_001079811.2 | NP_001073279.1 | 11, 12 | 1175 | |||
NM_001135602.2 | NP_001129074.1 | 8, 9 | 888 | |||
NM_001317040.1 | NP_001303969.1 | 12, 13 | 1425 | |||
GenBank mRNA | AK130133.1 | - | 11, 12 | 1209 | ||
AK222763.1 | - | 11, 12 | 1172 | |||
AK223091.1 | - | 11, 12 | 1197 | |||
AK298985.1 | - | 8, 9 | 800 | |||
AK300021.1 | - | 11, 12 | 1108 | |||
AK300753.1 | - | 12, 13 | 1341 | |||
AK312988.1 | - | 11, 12 | 1168 | |||
BC007493.2 | - | 11, 12 | 1183 | 2988086 | ||
BT007147.1 | - | 11, 12 | 1136 | |||
M22590.1 | - | 11, 12 | 1170 | |||
M27507.1 | - | 11, 12 | 1186 | |||
M27508.1 | - | 8, 9 | 793 | |||
M34423.1 | - | 11, 12 | 1196 |