Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dach2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142570.1 | NP_001136042.1 | 1 | 513 | ||
NM_033605.2 | NP_291083.1 | 1 | 513 | |||
GenBank mRNA | AF257217.1 | - | 1 | 512 | ||
AK045154.1 | - | 1 | 516 | |||
BC059233.1 | - | 1 | 502 | 6406445 |