Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ophn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052976.3 | NP_443208.1 | 4 | 896 | ||
GenBank mRNA | AK028822.1 | - | 4 | 380 | ||
AK031419.1 | - | 4 | 889 | |||
AK036038.1 | - | 4 | 897 | |||
AK039304.1 | - | 3 | 1064 | |||
AK043239.1 | - | 4 | 897 | |||
AK046214.1 | - | 4 | 352 | |||
AK048153.1 | - | 4 | 897 | |||
AK082551.1 | - | 4 | 536 | |||
AK087469.1 | - | 4 | 360 | |||
BC004845.1 | - | 5 | 494 | 2650009 |