Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rheb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053075.3 | NP_444305.2 | 6 | 496 | ||
GenBank mRNA | AB039919.1 | - | 6 | 546 | ||
AK151213.1 | - | 6 | 528 | |||
AY197373.1 | - | 6 | 496 | |||
BC012273.1 | - | 6 | 513 | 3710454 |