Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kars | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130868.2 | NP_001124340.1 | 3 | 472 | ||
NM_001286384.1 | NP_001273313.1 | 4 | 530 | |||
NM_053092.3 | NP_444322.1 | 2 | 292 | |||
GenBank mRNA | AK089053.1 | - | 2 | 217 | ||
AK090203.1 | - | 2 | 217 | |||
AK153236.1 | - | 2 | 215 | |||
AK167693.1 | - | 2 | 217 | |||
BC027356.1 | - | 4 | 442 | 4950333 | ||
BC035324.1 | - | 3 | 384 | 2811286 | ||
BC036289.1 | - | 2 | 261 | 3157706 | ||
BY792549.1 | - | 2 | 243 |