Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm11128 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001201389.1 | NP_001188318.1 | 3 | 742 | ||
GenBank mRNA | AF163316.1 | - | 3 | 741 |
Gene Symbol: Cml3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037842.3 | NP_001032931.1 | 3 | 1175 | ||
GenBank mRNA | AK046314.1 | - | 3 | 1176 | ||
BC014696.1 | - | 913 | 4237604 |
Gene Symbol: Gm4477 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253910.2 | NP_001240839.2 | 610 | |||
GenBank mRNA | AB221545.1 | - | 1 | 489 | ||
BC092089.1 | - | 603 | 4975242 |