Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nrep | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109988.1 | NP_001103458.1 | 5 | 547 | ||
NM_001109989.2 | NP_001103459.1 | 5 | 470 | |||
NM_001267717.1 | NP_001254646.1 | 5 | 459 | |||
NM_053078.4 | NP_444308.1 | 4 | 585 | |||
GenBank mRNA | AK020629.1 | - | 5 | 408 | ||
AK048361.1 | - | 4 | 385 | |||
AK081502.1 | - | 4 | 386 | |||
AK133769.1 | - | 3 | 1680 | |||
AK201102.1 | - | 1 | 113 | |||
BC054762.1 | - | 4 | 367 | 6414312 | ||
BQ715224.1 | - | 4 | 372 | |||
BU057200.1 | - | 4 | 375 | |||
CX768079.1 | - | 5 | 468 | |||
EU447302.1 | - | 3 | 313 | |||
X70398.1 | - | 4 | 381 |