Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lmod1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053106.2 | NP_444336.2 | 2 | 991 | ||
GenBank mRNA | AK079173.1 | - | 2 | 992 | ||
AK133276.1 | - | 2 | 994 |