Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smarca1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290708.1 | NP_001277637.1 | 2 | 392 | ||
NM_053123.4 | NP_444353.3 | 2 | 392 | |||
GenBank mRNA | AF325920.1 | - | 2 | 399 | ||
AK030741.1 | - | 2 | 359 | |||
AK032978.1 | - | 2 | 360 | |||
AK035068.1 | - | 2 | 272 | |||
AK038470.1 | - | 3 | 311 | |||
AK052092.1 | - | 2 | 363 | |||
BC016071.1 | - | 2 | 369 | |||
BC057115.1 | - | 2 | 392 | 6406330 | ||
BU704042.1 | - | 2 | 392 |