Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAFAH1B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000430.3 | NP_000421.1 | 11 | 2528 | ||
XM_011523901.1 | XP_011522203.1 | 12 | 2577 | |||
XM_011523902.2 | XP_011522204.1 | 12 | 2409 | |||
XM_011523903.2 | XP_011522205.1 | 12 | 2327 | |||
XM_017024699.1 | XP_016880188.1 | 11 | 2278 | |||
XM_017024700.1 | XP_016880189.1 | 11 | 2355 | |||
XM_017024701.1 | XP_016880190.1 | 11 | 2271 | |||
XM_017024702.1 | XP_016880191.1 | 10 | 2116 | |||
GenBank mRNA | BC064638.1 | - | 11 | 2170 | 5786560 | |
L13385.1 | - | 11 | 2177 | |||
L13387.1 | - | 2397 |