Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053183.2 | NP_444413.1 | 1, 2 | 170 | ||
GenBank mRNA | AF334104.1 | - | 1, 2 | 171 | ||
BC049906.1 | - | 1, 2 | 172 | |||
BC055440.1 | - | 1, 2 | 164 | |||
BC075714.1 | - | 1, 2 | 122 | |||
BC090996.1 | - | 1, 2 | 190 | 6757408 |