Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Col4a6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053185.2 | NP_444415.2 | 4 | 458 | ||
GenBank mRNA | AB041351.1 | - | 4 | 458 | ||
AK165178.1 | - | 4 | 440 | |||
BC057648.1 | - | 4 | 356 | 3500535 | ||
CF538953.1 | - | 4 | 455 |