Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OXCT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000436.3 | NP_000427.1 | 17 | 2990 | ||
XM_017009494.1 | XP_016864983.1 | 16 | 2781 | |||
GenBank mRNA | AK125115.1 | - | 1 | 3566 | ||
BC009001.1 | - | 17 | 2714 | 4148643 | ||
U62961.1 | - | 17 | 2757 |