Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ric8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053194.4 | NP_444424.1 | 3 | 579 | ||
GenBank mRNA | AF288813.1 | - | 3 | 578 | ||
AK031466.1 | - | 3 | 526 | |||
AK080192.1 | - | 3 | 522 | |||
AK155282.1 | - | 3 | 554 | |||
AK165536.1 | - | 3 | 556 | |||
AK167746.1 | - | 3 | 582 | |||
AK169367.1 | - | 3 | 543 | |||
AK171093.1 | - | 1 | 871 | |||
AK183673.1 | - | 1 | 73 | |||
AK187026.1 | - | 1 | 52 | |||
AK188871.1 | - | 1 | 250 | |||
BC022917.1 | - | 3 | 556 | 4460521 |