Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Egln1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053207.2 | NP_444437.2 | 2, 3 | 1150 | ||
GenBank mRNA | AF453878.1 | - | 2, 3 | 339 | ||
AJ310546.1 | - | 2, 3 | 853 | |||
BC006903.1 | - | 2, 3 | 150 | 3600175 | ||
BC083146.1 | - | 2, 3 | 457 |