Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cldn16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053241.5 | NP_444471.1 | 1, 2 | 473 | ||
GenBank mRNA | AB009016.1 | - | 1, 2 | 135 | ||
AF323748.1 | - | 1, 2 | 263 | |||
AK085268.1 | - | 1, 2 | 474 | |||
AK085333.1 | - | 1, 2 | 449 | |||
BC115574.2 | - | 2, 3 | 252 | 40102178 | ||
BC115575.2 | - | 2, 3 | 252 | 40102179 |