Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcdhb12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053137.2 | NP_444367.1 | 1 | 1096 | ||
GenBank mRNA | AY013773.1 | - | 1 | 914 | ||
BC127609.1 | - | 1 | 914 | 40059392 | ||
BC129838.1 | - | 1 | 914 | 40059393 |