Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hrg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053176.2 | NP_444406.2 | 2 | 350 | ||
GenBank mRNA | AB055897.1 | - | 2 | 308 | ||
AF194028.1 | - | 2 | 290 | |||
AY135662.1 | - | 2 | 311 | |||
BC011168.1 | - | 2 | 296 | 4195766 | ||
CB593922.1 | - | 2 | 322 |