Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cldn16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053241.5 | NP_444471.1 | 3 | 720 | ||
GenBank mRNA | AB009016.1 | - | 3 | 382 | ||
AF323748.1 | - | 3 | 510 | |||
AK085268.1 | - | 3 | 721 | |||
AK085333.1 | - | 3 | 696 | |||
BC115574.2 | - | 4 | 499 | 40102178 | ||
BC115575.2 | - | 4 | 499 | 40102179 |