Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Foxp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286607.1 | NP_001273536.1 | 8 | 1318 | ||
NM_053242.4 | NP_444472.2 | 10 | 1718 | |||
NM_212435.1 | NP_997600.1 | 8 | 1381 | |||
GenBank mRNA | AF339106.1 | - | 7 | 1059 | ||
AK051668.1 | - | 10 | 1701 | |||
AK082361.1 | - | 10 | 1390 | |||
AY079003.1 | - | 10 | 1347 | |||
BC058960.1 | - | 8 | 1315 | 6811877 | ||
BC062926.1 | - | 8 | 1381 | 6851153 |