Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcdhb20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053145.2 | NP_444375.2 | 1 | 2938 | ||
GenBank mRNA | AF326313.1 | - | 1 | 2914 | ||
AK045399.1 | - | 1 | 2939 | |||
AK045527.1 | - | 1 | 2911 | |||
AK046137.1 | - | 1 | 2931 | |||
BC031895.1 | - | 1 | 2873 | 5364116 |