Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IRAK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007199.2 | NP_009130.2 | 2 | 303 | ||
GenBank mRNA | AF113136.1 | - | 2 | 264 | ||
AK314040.1 | - | 2 | 285 | |||
BC057800.1 | - | 2 | 278 | 30335802 | ||
BC069388.1 | - | 2 | 251 | 7262303 | ||
BP292474.1 | - | 1 | 234 |