Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hrg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053176.2 | NP_444406.2 | 7 | 972 | ||
GenBank mRNA | AB055897.1 | - | 7 | 930 | ||
AF194028.1 | - | 7 | 912 | |||
AY135662.1 | - | 7 | 933 | |||
BC011168.1 | - | 7 | 918 | 4195766 | ||
CB946752.1 | - | 3 | 320 |