Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UGT1A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000463.2 | NP_000454.1 | 1 | 277 | ||
GenBank mRNA | AK290834.1 | - | 1 | 281 | ||
AY435136.1 | - | 1 | 262 | |||
BC128414.1 | - | 1 | 278 | 40117598 | ||
BC128415.1 | - | 1 | 277 | 40117599 | ||
DQ364247.1 | - | 1 | 262 | |||
JQ686667.1 | - | 1 | 262 | |||
JQ699637.1 | - | 1 | 262 | |||
JQ699638.1 | - | 1 | 262 | |||
JQ699639.1 | - | 1 | 262 | |||
JQ699640.1 | - | 1 | 262 | |||
JQ699641.1 | - | 1 | 262 | |||
JQ699642.1 | - | 1 | 262 | |||
JQ699643.1 | - | 1 | 262 | |||
JQ699644.1 | - | 1 | 262 | |||
JQ699645.1 | - | 1 | 262 | |||
JQ699646.1 | - | 1 | 262 | |||
JQ699647.1 | - | 1 | 262 | |||
JQ699649.1 | - | 1 | 262 | |||
JQ699650.1 | - | 1 | 262 | |||
M57899.1 | - | 1 | 277 |