Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Foxp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286607.1 | NP_001273536.1 | 13 | 1832 | ||
NM_053242.4 | NP_444472.2 | 15 | 2232 | |||
NM_212435.1 | NP_997600.1 | 13 | 1895 | |||
GenBank mRNA | AF339106.1 | - | 12 | 1573 | ||
AK051668.1 | - | 15 | 2215 | |||
AK082361.1 | - | 15 | 1904 | |||
AY079003.1 | - | 15 | 1861 | |||
BC058960.1 | - | 13 | 1829 | 6811877 | ||
BC062926.1 | - | 13 | 1895 | 6851153 |