Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc5a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF235001.1 | - | 15 | 2226 | ||
BC137650.1 | - | 15 | 2195 | 8860669 | ||
BC137651.1 | - | 15 | 2195 | 8860670 |